Adaptor tri-clamp hose connector, BOLA
Lieferant: Bohlender
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
D915-02
BOHLD915-02EA 30.5 EUR InStock
BOHLD915-02
Adaptor tri-clamp hose connector, BOLA
Fittings und Verbinder
Fitting made of PTFE for transition of Tri-Clamp as per DIN 32 676 to a hose connector.
  • Temperature resistant from –200 to 250 °C
  • Universal chemical resistance
  • The flowing product is only exposed to PTFE
Used for connection of elastic tubing (such as silicone, Viton®, Tygon®).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge