Sealing tapes, flat
Lieferant: Bohlender
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
H959-34 H959-28 H959-22 H959-16
BOHLH959-34EA 186 EUR InStock
BOHLH959-34 BOHLH959-28 BOHLH959-22 BOHLH959-16
Sealing tapes, flat
Klebebänder
Deformable, virginal PTFE flat tape with expanded fibre structure. For making customized gaskets “on-site”. FDA conform.
  • Tasteless and not ageing
  • Temperature resistance from −200 to +270 °C
  • Self-adhesive and physiologically safe
  • Almost universal chemical resistance
  • Quick and easy assembly, good sealing also on uneven surfaces
Different lengths available.
Colour: White
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge