Screw caps for bottles with GL threads, BOLA
Lieferant: Bohlender
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
H986-04 H986-02 H986-03 H986-05 H986-01
BOHLH986-04EA 40.3 EUR InStock
BOHLH986-04 BOHLH986-02 BOHLH986-03 BOHLH986-05 BOHLH986-01
Screw caps for bottles with GL threads, BOLA
Verschlüsse Flaschenverschlüsse
Screw cap with handy knurl, made of PTFE.
  • Very good chemical resistance
  • Temperature resistant from −200 to +250 °C
Suitable for bottles and vessels with GL threads.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge