Screw caps for bottles with GL threads, BOLA
Lieferant: Bohlender
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
H987-05 H987-04 H987-03 H987-02 H987-01
BOHLH987-05EA 5.3 EUR InStock
BOHLH987-05 BOHLH987-04 BOHLH987-03 BOHLH987-02 BOHLH987-01
Screw caps for bottles with GL threads, BOLA
Verschlüsse Flaschenverschlüsse
Screw cap made of PBTP, red, with handy knurl and integrated PTFE/silicone gasket.
  • Very good chemical resistance
  • Temperature resistant from −50 to +140 °C
Suitable for bottles and vessels with GL threads.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge