Eco stopcock plugs, without bore and taper 1:10
Lieferant: Bohlender
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
J1200-18 J1200-29 J1200-21 J1200-14 J1200-12 J1200-10 J1200-24 J1240-14 J1250-18 J1240-21 J1240-18
BOHLJ1200-18EA 8 EUR InStock
BOHLJ1200-18 BOHLJ1200-29 BOHLJ1200-21 BOHLJ1200-14 BOHLJ1200-12 BOHLJ1200-10 BOHLJ1200-24 BOHLJ1240-14 BOHLJ1250-18 BOHLJ1240-21 BOHLJ1240-18
Eco stopcock plugs, without bore and taper 1:10
Hähne
Made of PFTE, white, with grip and nut of PP, red.
  • Universal chemical resistance
  • Temperature resistance: from −10 to +120 °C
  • Taper: 1:10
  • Without bore
  • Washer included
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge