Rods, filtering, BOLA
Lieferant: Bohlender
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
N1505-32 N1520-28 N1510-28 N1505-28
BOHLN1505-32EA 96 EUR InStock
BOHLN1505-32 BOHLN1520-28 BOHLN1510-28 BOHLN1505-28
Rods, filtering, BOLA
Filter Filterrohre
These filtering rods are made of microporous PTFE.
  • Resistant to chemicals
  • Temperature resistance from –200 to +250 °C
Diameter and length are nominal dimensions and can contain a machining tolerance.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge