Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Pressure compensation sets and membranes, BOLA

Lieferant: Bohlender

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

N1698-18 N1699-18 N1699-45 N1699-32 N1699-25 N1699-14 N1698-45 N1698-32 N1698-25 N1698-14
BOHLN1698-18EA 8 EUR InStock
BOHLN1698-18 BOHLN1699-18 BOHLN1699-45 BOHLN1699-32 BOHLN1699-25 BOHLN1699-14 BOHLN1698-45 BOHLN1698-32 BOHLN1698-25 BOHLN1698-14
Pressure compensation sets and membranes, BOLA
Ventile Überdruckventile
Pressure compensation sets consisting of screw cap made of PPS for GL thread, PTFE/silicone gasket and exchangeable filtering membrane made of PTFE. Filtering membranes (2 µm) made of PTFE.

  • Temperature resistance: from −200 to +230 °C
  • Universal chemical resistance
  • Vacuum suitable
  • Autoclavable: 121 °C

For pressure compensation during filling or drawing of liquids. Prevention of unintentional overpressure or vacuum in the vessel. Integrated membrane prevents contamination of the product.

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge