Schläuche, PFA
Lieferant: Bohlender
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
S1811-02 S1811-04 S1811-06 S1811-28 S1811-18 S1811-40 S1811-24 S1811-30 S1811-10 S1811-14 S1811-50 S1811-12 S1811-08 S1811-26 S1811-22 S1811-20 S1811-16 S1811-05 S1811-07 S1811-03
BOHLS1811-02EA 11.4 EUR InStock
BOHLS1811-02 BOHLS1811-04 228-1197 BOHLS1811-28 BOHLS1811-18 BOHLS1811-40 BOHLS1811-24 BOHLS1811-30 BOHLS1811-10 BOHLS1811-14 BOHLS1811-50 BOHLS1811-12 BOHLS1811-08 BOHLS1811-26 BOHLS1811-22 BOHLS1811-20 BOHLS1811-16 BOHLS1811-05 BOHLS1811-07 BOHLS1811-03
Schläuche, PFA
Schläuche Transferschlauch
Made of PFA, transparent
  • Temperature resistance: −200 to +260°C
  • Gastighttubing
  • Almost universal chemical resistance
  • Resistant to irradiation and weather
  • Sterilized in autoclaves
  • Mechanical strength even at high temperatures
Applications include perfect tubing for aggressive and pure liquids or gases. Good bending radius and burst pressure.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge