Desiccator cabinet, anti-static, SICCO Star
Lieferant: Bohlender
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
V1910-07
BOHLV1910-07EA 2160 EUR InStock
BOHLV1910-07
Desiccator cabinet, anti-static, SICCO Star
Exsikkatoren Trockenkammern
PC desiccator cabinet which has reinforced aluminium frame with antistatic acrylic panels and easily readable electronic hygrometer. Mainly used for temporary storage of components for electronic products.
  • Variable height telescopic shelves made of aluminium or stainless steel
  • Non-slip rubber feet or four casters (two of the casters with brakes)
  • Door with magnetic catch and circular rubber seal
  • Connection for grounding cable and tray for desiccant
These desiccators can protect electronic components against humidity and airborne particulate. Electrostatic charges are discharged by a grounding cable and a charge neutral atmosphere is produced inside the desiccator.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge