Desiccator cabinets, anti-static, SICCO maxi series
Lieferant: Bohlender
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
V1922-07 V1923-07
BOHLV1922-07EA 4660 EUR InStock
BOHLV1922-07 BOHLV1923-07
Desiccator cabinets, anti-static, SICCO maxi series
Exsikkatoren Trockenkammern
Desiccator cabinets, PC ESD, have a reinforced aluminium frame with panels made of antistatic polycarbonate and an easy to read electronic hygrometer. They are mainly used for temporary storage of components for electronic products.
  • Door with magnetic catch and circular rubber seal
  • Four casters (two of the casters with brakes)
  • Variable height telescopic shelves made of stainless steel
  • Connection for grounding cable and tray for desiccant
These desiccators can protect electronic components against humidity and airborne particulate. Electrostatic charges are discharged by a grounding cable and a charge neutral atmosphere is produced inside the desiccator.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge