Desiccator cabinets, SICCO maxi series
Lieferant: Bohlender
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
V1927-07
BOHLV1927-07EA 4020 EUR InStock
BOHLV1927-07
Desiccator cabinets, SICCO maxi series
Exsikkatoren Trockenkammern
Orange acrylic cabinets have a reinforced aluminium frame, door with foam rubber gasket and magnetic strip, orange acrylic panels reduce light incidence and protect against ultraviolet radiation. Especially suitable for storage of light-sensitive substances
  • Door mounted electronic hygrometer indicates minimum and maximum temperature and humidity values
  • Temperature resistance from −20 to +70 °C
  • Easy to move about as cabinets have four castors
Load capacity: 30 kg/shelf, 160 kg (total)
Load capacity: 30 kg/shelf, 160 kg (total)
Lieferumfang: Supplied with four removable, stainless steel, telescopic shelves and one desiccant tray. Capacity to accommodate maximum of 34 shelves. Silica gel not included.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge