Desiccator cabinet, SICCO Super Big Star
Lieferant: Bohlender
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
V1935-07
BOHLV1935-07EA 2140 EUR InStock
BOHLV1935-07
Desiccator cabinet, SICCO Super Big Star
Exsikkatoren Trockenkammern
SICCO desiccators are designed for storing and drying humidity sensitive products using silica gel. The tight fitting door protects the contents from contamination in the atmosphere.
  • Reinforced aluminium frame with acrylic panels
  • Door with magnetic catch and circular rubber seal
  • Non slip rubber feet
  • Easy to read electronic hygrometer indicates minimum and maximum temperature and humidity values
  • Variable height shelves made of stainless steel
  • Telescopic shelves
  • Temperature resistance: –20 to +70 °C
The controlled environment inside the desiccator makes it ideal for storing reference materials, retained samples, metallographic specimens and DNA samples.
Lieferumfang: Supplied with two stainless steel shelves, tray and hygrometer and hose (5 m) with two quick couplings for gas filling, usable with up to eight shelves.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge