Superblast Safety Goggles
Lieferant: Bollé Safety
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
SUPBLEPSI
BOLESUPBLEPSIEA 19.1 EUR InStock
BOLESUPBLEPSI
Superblast Safety Goggles
Vollsichtbrillen
Superblast goggles gets softer frame, a platinum version and a simplified intuitive assembly system goggle-Visor.
  • Flexible and clear frame for increased comfort
  • Wide choice of frame types for all applications
  • Intuitive and simple goggle-visor assembly system
Lens: Polycarbonate (2 mm)
Curve: Slightly curved
Frame material: PVC
Strap: Nylon Polyster (250 mm)
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge