Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ADNP Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-0039R-CY7
BOSSBS-0039R-CY7EA 883 EUR InStock
BOSSBS-0039R-CY7
Anti-ADNP Rabbit Polyclonal Antibody (Cy7®)
Antikörper
The activity dependent neuroprotective protein (ADNP) gene is frequently amplified in many neoplasias, including breast, bladder, ovarian, pancreatic, and colon cancers. ADNP mRNA is abundantly expressed in distinct normal tissues, and high expression levels were encountered in malignant cells. ADNP is implicated in maintaining cell survival, perhaps through modulation of p53. The encoded protein contains one homeobox and nine zinc finger domains, suggesting that it functions as a transcription factor.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ADNP
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge