Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CASP4 + CASP 13 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-0053R-CY5.5
BOSSBS-0053R-CY5.5EA 900 EUR InStock
BOSSBS-0053R-CY5.5
Anti-CASP4 + CASP 13 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Caspase-13 (also designated Evolutionarily related interleukin-1beta converting enzyme ; ERICE) involved in the activation cascade of caspases responsible for apoptosis execution. Might function by either activating some proteins required for cell death or inactivating proteins necessary for cell survival. [subunit] Heterodimer of a small and a large subunit. The two subunits are derived from the precursor sequence by an autocatalytic mechanism or by cleavage by Caspase-8. Mainly expressed in peripheral blood lymphocytes, spleen and placenta. Belongs to peptidase family C14.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CASP4 + CASP 13
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Bovine

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge