Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-RTN4R Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-0129R-A488
BOSSBS-0129R-A488EA 893 EUR InStock
BOSSBS-0129R-A488
Anti-RTN4R Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Receptor for RTN4, OMG and MAG. Mediates axonal growth inhibition and may play a role in regulating axonal regeneration and plasticity in the adult central nervous system (By similarity). Acts in conjunction with RTN4 and LIGO1 in regulating neuronal precursor cell motility during cortical development.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RTN4R
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge