Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-DEFB1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-0241R-HRP
BOSSBS-0241R-HRPEA 1350 EUR InStock
BOSSBS-0241R-HRP
Anti-DEFB1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
The attachment of enveloped viruses to cells and the fusion of viral and cellular membranes are critical early events in the HIV viral infection. This process is mediated by envelope glycoproteins (gp) on the surface of the virus. The human immunodeficiency virus type 1 (HIV-1) envelope glycoprotein, gp160, is proteolytically cleaved into gp120 and gp41, which remain noncovalently associated with one another. gp120 is one of the proteins that forms the envelope of HIV. gp120 projects from the surface of HIV and binds to the CD4 molecule on helper T cells. gp120 has been a logical experimental HIV vaccine because the outer envelope is the first part of the virus that encounters antibody. gp41 is embedded in the outer envelope of HIV that anchors gp120. gp41 also plays a key role in HIV's infection of CD4+ T cells by facilitating the fusion of the viral and cell membranes. The nomenclature of the gp proteins describes their respective molecular masses (e.g., gp160, gp120, gp41).

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: DEFB1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge