Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-EXPB2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-0243R-A647
BOSSBS-0243R-A647EA 893 EUR InStock
BOSSBS-0243R-A647
Anti-EXPB2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
beta-expansin (EXBP2) may cause loosening and extension of plant cell walls by disrupting noncovalent bonding between cellulose microfibrils and matrix glucans. No enzymatic activity has been found Subcellular Location at Cell-wall bound. Belongs to the expansin family.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: EXPB2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Plant

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge