Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Neurobeachin Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-0268R-CY3
BOSSBS-0268R-CY3EA 900 EUR InStock
BOSSBS-0268R-CY3
Anti-Neurobeachin Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Neurobeachin binds to type II regulatory subunits of protein kinase A and anchors/targets them to the membrane. It may anchor the kinase to cytoskeletal and/or organelle-associated proteins. Neurobeachin, is also a neuron-specific multidomain protein of 327 kDa with a high-affinity binding site for the type II regulatory subunit of protein kinase A. Neurobeachin is peripherally associated with pleomorphic tubulovesicular endomembranes near the trans sides of Golgi stacks and throughout the cell body and cell processes. It is also found in a subpopulation of synapses, where it is concentrated at the postsynaptic plasma membrane.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Neurobeachin
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Other Species

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge