Anti-PBEF CT Rabbit Polyclonal Antibody (Cy5.5®)
Lieferant: Bioss
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BS-0273R-CY5.5
BOSSBS-0273R-CY5.5EA 900 EUR InStock
BOSSBS-0273R-CY5.5
Anti-PBEF CT Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
icotinamide phosphoribosyltransferase (NAmPRTase) (Nampt) (Pre-B cell-enhancing factor) (Pre-B-cell colony-enhancing factor 1) (Fisfatin) Catalyzes the condensation of nicotinamide with5-phosphoribosyl-1-pyrophosphate to yield nicotinamide mononucleotide, an intermediate in the biosynthesis of NAD. It is the rate limiting component in the mammalian NAD biosynthesis pathway. Expressed in large amounts in bone marrow, liver tissue, and muscle. Also present in heart, placenta, lung, and kidney tissues. Subcellular Location is in Cytoplasm. Belongs to the NAPRTase family.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PBEF CT
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge