Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SSB Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-0276R-CY3
BOSSBS-0276R-CY3EA 900 EUR InStock
BOSSBS-0276R-CY3
Anti-SSB Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Lupus La protein (Sjogren syndrome type B antigen) (SS-B) (La ribonucleoprotein) (La autoantigen) plays a role in the transcription of RNA polymerase III. It is most probably a transcription termination factor. Binds to the 3' termini of virtually all nascent polymerase III transcripts. It is associated with precursor forms of RNA polymerase III transcripts including tRNA and 4.5S, 5S, 7S, and 7-2 RNAs. The phosphorylation sites are at the C-terminal part of the protein. Sera from patients with systemic lupus erythematosus(SLE) often contain that react with the normal cellular La protein as if this antigen was foreign.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SSB
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge