Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-FGL2 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-0315R-CY5.5
BOSSBS-0315R-CY5.5EA 883 EUR InStock
BOSSBS-0315R-CY5.5
Anti-FGL2 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
FGL2 is a secreted protein that is similar to the beta- and gamma-chains of fibrinogen. The carboxyl-terminus of the encoded protein consists of the fibrinogen-related domains (FRED). The encoded protein forms a tetrameric complex which is stabilized by interchain disulfide bonds. It may play a role in physiologic functions at mucosal sites. It is constitutively expressed in cytotoxic T-cells. Lack of expression in other lymphoid- and nonlymphoid-derived cell lines suggested that expression of FGL2 may be restricted to lymphocytes. FGL2 is induced via a mechanism involving IFNG and components of the IFNG signaling pathway, including STAT1 and IRF1.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FGL2
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge