Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-E1 glycoProtein Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-0319R-A680
BOSSBS-0319R-A680EA 890 EUR InStock
BOSSBS-0319R-A680
Anti-E1 glycoProtein Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Rubella virus (RV), the sole member of the genus Rubivirus within the family Togaviridae, is a small enveloped, positive strand RNA virus. The nucleocapsid consists of 40S genomic RNA and a single species of capsid protein which is enveloped within a host-derived lipid bilayer containing two viral glycoproteins, E1 (58 kDa) and E2 (42-46 kDa). In virus infected cells, RV matures by budding either at the plasma membrane, or at the internal membranes depending on the cell type and enters adjacent uninfected cells by a membrane fusion process in the endosome, directed by E1-E2 heterodimers. The heterodimer formation is crucial for E1 transport out of the endoplasmic reticulum to the Golgi and plasma membrane. In RV E1, a cysteine at position 82 is crucial for the E1-E2 heterodimer formation and cell surface expression of the two proteins. E1 has been shown to be a type 1 membrane protein, rich in cysteine residues with extensive intramolecular disulphide bonds [PMID: 11682134].

Type: Primary
Antigen: E1 glycoprotein
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 801-900/1063
Host: Rabbit
Isotype: IgG
Reactivity: Virus

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge