Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PIP5KL1 Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-0451R
BOSSBS-0451REA 725 EUR InStock
BOSSBS-0451R
Anti-PIP5KL1 Rabbit Polyclonal Antibody
Antikörper
PIPKH, also known as PIP5KL1 (phosphatidylinositol-4-phosphate 5-kinase-like 1), is a 394 amino acid phosphoinositide kinase-like protein that contains one PIPK domain. Although PIPKH lacks intrinsic lipid kinase activity, it associates with type I PIPKs and may play a role in localization of PIPK activity. Encoded by a gene that maps to human chromosome 9q34.11, PIPKH localizes to cytoplasm, specifically to large cytoplasmic vesicular structures, and exists as two alternatively spliced isoforms. Highly expressed in brain and testis, PIPKH is also expressed at very low levels in heart, spleen, lung, liver, skeletal muscle and kidney. PIPKH heterodimerizes with other type I phosphatidylinositol-4-phosphate 5-kinases, and may function as a scaffold to localize and regulate kinases to specific cell compartments. Overexpression of PIPKH may suppress cell proliferation and migration in human gastric cancer cells and may also inhibit cervical cancer formation.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: PIP5KL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge