Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SPINT1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-0540R-A647
BOSSBS-0540R-A647EA 893 EUR InStock
BOSSBS-0540R-A647
Anti-SPINT1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
HGFA Inhibitor 1 is an endogenous inhibitor of the kunitz-type serine proteinase HGF-activator (HGFA). HGF was first described as a hepatocyte-specific mitogen and survival factor, and has since been shown to exert a variety of actions on many cell types by binding to its MET receptor. HGF is activated by cleavage of the single-chain form to form a two-chain version by HGFA. HGFA Inhibitor 1 also inhibits trypsin, and MTSP-1 (Matriptase), a serine proteinase that also activates HGF. HGFB Inhibitor 1 is a splice variant of HGFA Inhibitor 1, and contains a 16 amino acid insert after the first kringle domain, relative to the HGFA Inhibitor 1 sequence.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SPINT1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge