Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-DPPA2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-0597R-HRP
BOSSBS-0597R-HRPEA 900 EUR InStock
BOSSBS-0597R-HRP
Anti-DPPA2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
SCRG1 (scrapie-responsive protein 1) is a 98 amino acid secreted protein that belongs to the SCRG1 family. The SCRG1 protein contains a 20-amino acid signal peptide, and is expressed abundantly in the central nervous system of human adult, but not at all in fetal brain. The protein is targeted to the Golgi apparatus and large dense-core vesicles/secretory granules in neurons. High levels of SCRG1 transcripts are also observed in testis and aorta. SCRG1 is associated with neurodegenerative changes observed in transmissible spongiform encephalopathies. It may play a role in host response to prion-associated infections. The SCRG1 protein may be partly included in the membrane or secreted by the cells due to its hydrophobic N-terminus. The human and mouse SCRG1 proteins share 83% sequence identity. The SCRG1 gene is conserved in chimpanzee, cow, mouse, rat and chicken, and maps to human chromosome 4q34.1.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: DPPA2
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge