Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Tyrosinase Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-0819R-A680
BOSSBS-0819R-A680EA 890 EUR InStock
BOSSBS-0819R-A680
Anti-Tyrosinase Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Tyrosinase is the key enzyme for melanin,synthesis in mammalian melanocytes and has been considered to be a unique marker for the study of melanocyte differentiation. A cDNA library was constructed from poly(A)+ mRNA from mouse melanocytes and screened using anti- tyrosinase antiserum and oligonucleotide probes corresponding to amino acid sequence of tyrosinase. sequencing of some cDNA clones positive in these screenings gave a nucleotide sequence of 1838 nucleotides including a open reading frame of 1344 nucleotides that was found to correspond exactly to the amino acid sequence of the cyanogen bromide fragments of tyrosinase.

Type: Primary
Antigen: Tyrosinase
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 155-250/529
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge