Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-AMACR Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-0840R-CY7
BOSSBS-0840R-CY7EA 893 EUR InStock
BOSSBS-0840R-CY7
Anti-AMACR Rabbit Polyclonal Antibody (Cy7®)
Antikörper
alpha-methylacyl-CoA racemase(AMACR/P504S) is Prostate-specific antigen (PSA) screening for prostate cancer is now widespread in the United States among men of all ages. However PSA has limited specificity because benign disease, including prostatic enlargement and inflammation, can increase PSA levels. Thus, a more specific prostate cancer markers is needed. One such potential marker is AMACR, an enzyme that is involved in peroxisomal beta-oxidation of dietary branched-chain fatty acids. Recent studies have shown that, compared with expression in normal or benign prostate epithelium, AMACR is consistently overexpressed in prostate cancer epithelium, making it a specific marker for cancer cells within the prostate gland. Furthermore, overexpression of AMACR may increase the risk of prostate cancer because its expression is increased in premalignant lesions (prostatic intraepithelial neoplasia).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: AMACR
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge