Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-HP1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-0930R-FITC
BOSSBS-0930R-FITCEA 929 EUR InStock
BOSSBS-0930R-FITC
Anti-HP1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
HP1 alpha is a component of heterochromatin. It recognizes and binds histone H3 tails methylated at Lys-9, leading to epigenetic repression. HP1 alpha may interact with lamin B receptor (LBR). This interaction can contribute to the association of the heterochromatin with the inner nuclear membrane. HP1 proteins are relatively small proteins (~25 kDa) with a conserved amino-terminal chromo domain and a structurally related carboxy-terminal motif, the chromo shadow domain. Both domains of HP1 are required for binding to native chromatin in vivo, but they contribute differentially to binding in euchromatin and heterochromatin.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HP1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Other Species

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge