Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Acetyl-Histone H1 Lys52 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-0931R-A750
BOSSBS-0931R-A750EA 917 EUR InStock
BOSSBS-0931R-A750
Anti-Acetyl-Histone H1 Lys52 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Histone H1 protein binds to linker DNA between nucleosomes forming the macromolecular structure known as the chromatin fibre. Histones H1 are necessary for the condensation of nucleosome chains into higher-order structured fibres. Acts also as a regulator of individual gene transcription through chromatin remodelling, nucleosome spacing and DNA methylation (By similarity).

Type: Primary
Antigen: Acetyl-Histone H1 Lys52
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge