Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-TRIM32 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-0964R-CY7
BOSSBS-0964R-CY7EA 900 EUR InStock
BOSSBS-0964R-CY7
Anti-TRIM32 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Tripartite motif-containing protein 32 (TRIM32) belongs to the tripartite motif (TRIM) protein family. TRIM32, like all TRIM proteins, contains a domain structure composed of a B-box, a RING-finger and a coiled-coil motif. Additionally, TRIM32 has six C-terminal NHL domains; it is expressed mainly in the skeletal muscle. The TRIM32 gene encodes an E3 ubiquitin ligase, a protein that attaches ubiquitin to a lysine residue on a target protein and acts in conjunction with ubiquitin-conjugating enzymes UbcH5a, UbcH5c and UbcH6. Mutations in the TRIM32 gene cause two forms of autosomal recessive muscular dystrophy designated limb girdle muscular dystrophy type 2H (LGMD2H) and sarcotubular myopathy (STM). TRIM32 mutations can also result in Bardet-Biedl syndrome (BBS), an autosomal recessive disorder characterized by pigmentary retinopathy, polydactyly, hypogenitalism, renal abnormalities, learning disabilities and obesity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TRIM32
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge