Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-UEA1 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-10025R-CY5
BOSSBS-10025R-CY5EA 900 EUR InStock
BOSSBS-10025R-CY5
Anti-UEA1 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Ulex Europaeus is a European gorse shrub with fragrant golden-yellow flowers. Ulex Europaeus-I Lectin is a 46 kDa glycoprotein known to interact with -L fucosyl residues in oligosaccharides present on the membranes of human blood group O erythrocytes, human endothelial cells and a variety of human and animal epithelial cells.This antibody reacts with Ulex Europaeus Lectin 1 bound to human endothelial cells of normal and neoplastic blood and lymphatic vesicles. It also reacts with human epithelia such as in the colon, bronchus, epidermis, sweat gland ducts and hair follicles. It also reacts with for Ulex Europaeus Lectin 1 bound to vasclular endothelium and squamous epithelium of human tonsil. While erythrocytes may be stained, no other tonsilar elements are reactive with the antibody.This antibody may be useful in the detection of vascular tumors, the This antibody may be useful in the detection of vascular tumors, the investigation of vascular invasion by tumor cells, for determination of Ulex Europaeus Lectin 1 binding to normal, embryonal, dysplastic and neoplastic epithelia and the study of storage diseases such as fucosidosis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: UEA1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Plant

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge