Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-LAT Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-10126R-A555
BOSSBS-10126R-A555EA 893 EUR InStock
BOSSBS-10126R-A555
Anti-LAT Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Linker for activation of T cells (LAT) is an integral membrane protein (36 to 38 kD), which plays an important role in linking engagement of the TCR to the biochemical events of T cell activation. LAT antibody stains thymus and peripheral lymphoid tissues such as T cell areas in lymph nodes and spleen. LAT is expressed in T lymphocytes in interstitial spaces, platelets and megakaryocytes. LAT is not expressed in B cells, macrophages, plasma cells, monocytes, epithelial histiocytes and dendritic cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LAT
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge