Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-H3N1 hemagglutinin Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-10320R-FITC
BOSSBS-10320R-FITCEA 900 EUR InStock
BOSSBS-10320R-FITC
Anti-H3N1 hemagglutinin Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Hemagglutinin (HA) is a class I viral fusion protein from Influenza virus. It is a major glycoprotein, comprising over 80% of the envelope proteins present in the virus particle. HA binds to sialic acid-containing receptors on the cell surface, bringing about the attachment of the virus particle to the cell, and is responsible for penetration of the virus into the cell cytoplasm by mediating the fusion of the membrane of the endocytosed virus particle with the endosomal membrane. The extent of infection into host organism is determined by HA. In natural infection, inactive HA is matured into HA1 and HA2 outside the cell by one or more trypsin-like, arginine-specific endoproteases secreted by the bronchial epithelial cells. The HA protein is a homotrimer of disulfide-linked HA1-HA2. It also plays a major role in the determination of host range restriction and virulence. Genetic variation of hemagglutinin and/or neuraminidase genes results in the emergence of new influenza strains.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: H3N1 hemagglutinin
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge