Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-VGF/NERP2 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-10332R-CY7
BOSSBS-10332R-CY7EA 900 EUR InStock
BOSSBS-10332R-CY7
Anti-VGF/NERP2 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
This gene is specifically expressed in a subpopulation of neuroendocrine cells, and is upregulated by nerve growth factor. The structural organization of this gene is similar to that of the rat gene, and both the translated and the untranslated regions show a high degree of sequence similarity to the rat gene. The encoded secretory protein also shares similarities with the secretogranin/chromogranin family, however, its exact function is not known. Nerve growth factor (NGF) is a peptide that plays a key role in the differentiation and survival of neurons in the peripheral nervous system (PNS) and the central nervous system (CNS). VGF is a peptide synthesized and secreted by neurons and is upregulated by NGF in the PC12 cell line. VGF is widely expressed in both the PNS and CNS, but is especially abundant in the adult hypothalamus. VGF plays an essential role in how the brain regulates energy expenditure and body weight. Its expression is rapidly induced by injury, the circadian clock, and neuronal activity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: VGF/NERP2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge