Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SYNGAP Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-10392R-HRP
BOSSBS-10392R-HRPEA 811 EUR InStock
BOSSBS-10392R-HRP
Anti-SYNGAP Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
The PSD-95/SAP 90 family of proteins, which are known to bind to and cluster various membrane proteins, are involved in the organization of synaptic structure. SynGAP (for synaptic GTPase activating protein), a novel synaptic RasGAP, is a brain-specific protein abundant in the cortex, hippocampus and olfactory bulb. SynGAP interacts with all three PDZ domains within PSD-95/ SAP 90 proteins. It represents one of three classes of mammalian RasGAPs and is specifically localized to excitatory synapses possessing NMDA receptors. SynGAP may be involved in the regulation of BDNF as well as Ras signaling. Its activity is inhibited by phosphorylation by CaMKII, which could result in the activation of the MAP kinase pathway.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: SYNGAP
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge