Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-RARRES2 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-10410R-CY7
BOSSBS-10410R-CY7EA 883 EUR InStock
BOSSBS-10410R-CY7
Anti-RARRES2 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Chemerin, also known as retinoic acid receptor responder 2 (RARRES2) and tazarotene induced gene 2 (TIG2), is a chemoattractant protein for cells expressing G-protein-linked receptor chemokine-like receptor 1 (CMKLR1, ChemR23 or GPCR-DEZ). The protein is synthesized as a secreted precursor, prochemerin (18 kDa), which is poorly active. Upon proteolytic removal of the last six amino acids, prochemerin is converted into mature active chemerin (15-16 kDa) that acts as a strong and highly specific agonist of its receptor CMKLR1. Chemerin is a newly described adipokine with effects on adipocyte differnetiation and metabolism.

Recommended Dilutions: IF(IHC-P): 1:50-200; IF(ICC)(1:50-200

Type: Primary
Antigen: RARRES2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge