Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SEB Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-10722R-FITC
BOSSBS-10722R-FITCEA 883 EUR InStock
BOSSBS-10722R-FITC
Anti-SEB Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Staphylococcal enterotoxins represent a group of proteins, which are secreted by Staphylococcus aureus and cause staphylococcal food poisoning syndrome. The illness is characterised by high fever, hypotension, diarrhea, shock, and in some cases death. Their molecular masses range between 27 and 30 kDa. At present, seven enterotoxins are known, namely A, B, C1, C2, C3, D and E. Their amino acid sequences have been determined and it was shown that all are single chain polypeptides containing one disulfide bond formed by two half cystines located in the middle of the polypeptide chain, which form the so called cysteine loop. SEB is an extremely potent activator of T cells, stimulating the production and secretion of various cytokines which mediate many of the toxic effects of SEB.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Staphylococcus Enterotoxin B
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bacteria

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge