Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-NPTX2 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11017R-CY3
BOSSBS-11017R-CY3EA 900 EUR InStock
BOSSBS-11017R-CY3
Anti-NPTX2 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Long pentraxins are a family of highly conserved proteins that are expressed in the brain and central nervous system, and form multimeric complexes. Neuronal pentraxin 1 (NP1), NP2, and neuronal pentraxin receptor (NPR) are members of the long pentraxins that represent a neuronal uptake pathway that may function during synapse formation and remodeling. The NP1 gene is located on chromosome 17q25.3 and the protein product mediates the uptake of synaptic material, including the presynaptic snake venom toxin, taipoxin (3). NP2, whose function is unknown, is located on chromosome 7q22.1 and like NP1 contains several potential N-linked glycosylation sites. NPR is expressed on the cell membrane and can form heteropentamers with NP1 and NP2 that can be released from the cell membrane by proteolysis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NPTX2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge