Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GAN Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11025R-A555
BOSSBS-11025R-A555EA 893 EUR InStock
BOSSBS-11025R-A555
Anti-GAN Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Gigaxonin, also refered to as giant axonal neuropathy, GAN1, or KLHL16, controls protein degradation and is essential for neuronal function and survival. Gigaxonin is a member of the cytoskeletal BTB/kelch repeat family and influences cytoskeletal organization and dynamics, playing a large role in neurofilament architecture. The amino terminal BTB domain of gigaxonin binds to the ubiquitin-activating enzyme E1, while the carboxy-terminal kelch repeat domain interacts directly with the light chain of microtubule-associated protein 1B (MAP1B), and tags it for degredation. Overexpression of MAP1B may lead to neuronal cell death, whereas a reduction of MAP1B significantly improves the survival rate of neurons. Mutations in the Gigaxonin gene result in human giant axonal neuropathy (GAN), an autosomal recessive neurodegenerative disorder characterized by axonal degeneration caused by cytoskeletal abnormalities, including accumulated intermediate filaments.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Gigaxonin
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge