Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SETBP1 Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11029R
BOSSBS-11029REA 725 EUR InStock
BOSSBS-11029R
Anti-SETBP1 Rabbit Polyclonal Antibody
Antikörper
Midline-2 is a 715 amino acid protein encoded by the human gene MID2. Midline-2 belongs to the TRIM/RBCC family and contains two B box-type zinc fingers, one B30.2/SPRY domain, one COS domain, one fibronectin type-III domain and one RING-type zinc finger. Midline-2 is a cytoplasmic protein found as a homodimer or heterodimer with Midline-1. It also interacts with IGBP1 (Lymphocyte signaling protein A4). Dimerization is mediated by the tripartite motif (RBCC; RING- and B box-type zinc fingers and coiled coil domains) and microtubule association is dependent on the C-terminal B30.2 domain. Midline-2 is expressed at low levels in fetal kidney and lung, and in adult prostate, ovary and small intestine.

Recommended Dilutions: Western Blot: 1:100 to 1000; IHC-P: 1:100 to 500; IF(IHC-P): 1:50 to 200.

Type: Primary
Antigen: SETBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge