Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GLDN Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11032R-CY7
BOSSBS-11032R-CY7EA 900 EUR InStock
BOSSBS-11032R-CY7
Anti-GLDN Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Gliomedin is a 551 amino acid protein encoded by the human gene GLDN. Gliomedin is thought to play a role in the formation of the nodes of Ranvier along myelinated axons. Accumulation of Na+ channels at the nodes of Ranvier is a prerequisite for saltatory conduction. In peripheral nerves, clustering of these channels along the axolemma is regulated by myelinating Schwann cells through an unknown mechanism. Gliomedin is a glial ligand for Neurofascin and NrCAM, two axonal immunoglobulin cell adhesion molecules that are associated with Na+ channels at the nodes of Ranvier. Gliomedin is expressed by myelinating Schwann cells and accumulates at the edges of each myelin segment during development, where it aligns with the forming nodes. Gliomedin is a single-pass type II membrane protein localized to the nodes of Ranvier and is specifically expressed in spinal cord, brain, placenta and sciatic nerve. It is more abundant in peripheral than central nervous system.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Gliomedin
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge