Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-TANC1 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11037R-CY3
BOSSBS-11037R-CY3EA 900 EUR InStock
BOSSBS-11037R-CY3
Anti-TANC1 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
TANC (tetratricopeptide repeat, ankyrin repeat and coiled-coil domain-containing protein), also known as TANC1, is a 1,861 amino acid postsynaptic cell membrane protein that contains eleven ANK repeats, three TPR repeats and belongs to the TANC family. Considered a scaffolding component in the postsynaptic density, TANC interacts with TNIK, SAPAP1, Alpha-internexin, CaMKII, NMDA 2 and GluR-1. It is also thought that TANC interacts directly with SAP 97, PSD-95 and Homer. Upon stimulation by Rap 2, MINK1 and TNIK may phosphorylate TANC. The TANC gene encodes two alternatively spliced isoforms, contains approximately 264,025 bases and maps to human chromosome 2q24.2. Making up approximately 8% of the human genome, chromosome 2 consists of 237 million bases and encodes over 1,400 genes. A number of genetic diseases are linked to genes on chromosome 2 including Harlequin icthyosis, sitosterolemia and Alstré°‰ syndrome.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TANC1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge