Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-STXBP6 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11039R-A555
BOSSBS-11039R-A555EA 893 EUR InStock
BOSSBS-11039R-A555
Anti-STXBP6 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Amisyn is a mostly cytosolic protein related to Tomosyn which plays an important role in SNARE complex assembly. Amisyn contains a v-SNARE coiled coil homology domain that binds to Syntaxin 1A and weakly to Syntaxin 4. Three isoforms exist for Amisyn. Isoform 1 is the full length protein, isoform 2 has a different amino acid sequence between residues 204-210 and isoform 3 is missing amino acids 1-102 and contains a different sequence for amino acids 103-150. Amisyn lacks a transmembrane domain and therefore is unable to assemble into a functional, membrane-anchored SNARE complex. This suggests that Amisyn may instead be acting to maintain SNARE conformation and facilitate the binding of VAMP-2. Amisyn can inhibit exocytosis independent of Syntaxin binding.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: STXBP6
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge