Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-BEST2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11041R-A350
BOSSBS-11041R-A350EA 893 EUR InStock
BOSSBS-11041R-A350
Anti-BEST2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
The Bestrophins are a newly described family of anion channels unrelated in primary sequence to any previously characterized channel proteins. Bestrophins were originally defined as a family of over 20 related sequences of the C. elegans. The first mammalian Bestrophin was identified as the vitelliform macular dystrophy (VMD), 1 also known as Best disease. Three more members of the bestrophin family members were cloned and indentified recently, Bestrophin 2, 3 and 4. RT PCR analyses revealed tissue restricted expression of the three genes with both Bestrophin 1 and Bestrophin 2 are abundantly transcribed in colon. Functionally the bestrophines oligomerise to form tetramers and pentamers in order to act as calcium sensitive chloride channels. It has been shown that Bestrophin interacts with beta catalytic subunit of protein phosphatase 2A (PP2Ac). Such protein protein interaction between Bestrophin and PP2Ac and the structural subunit of PP2A, PR65, was confirmed by reciprocal immunoprecipitation. The interaction between PP2Ac and the Bestrophin takes place near the carboxy terminal end of the protein.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BEST2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge