Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-BEST4 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11043R-A647
BOSSBS-11043R-A647EA 893 EUR InStock
BOSSBS-11043R-A647
Anti-BEST4 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Bestrophin-4 is a 473 amino acid member of the Bestrophin family of proteins. Members of the Bestrophin family are transmembrane proteins that contain a high percentage of aromatic residues, a conserved RFP (Arg-Phe-Pro) motif and they function as anion channels. Bestrophin-4 acts as a calcium-sensitive chloride channel located in the cell membrane. It is believed that Bestrophin-4 also acts as a channel for other physiologically significant anions, such as bicarbonate. Bestrophin-4 is predominantly expressed in the colon, but can be found at low levels in testis, placenta, trachea, spinal chord, lung and retina.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BEST4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge