Anti-BEST4 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Lieferant: Bioss
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BS-11043R-HRP
BOSSBS-11043R-HRPEA 900 EUR InStock
BOSSBS-11043R-HRP
Anti-BEST4 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Bestrophin-4 is a 473 amino acid member of the Bestrophin family of proteins. Members of the Bestrophin family are transmembrane proteins that contain a high percentage of aromatic residues, a conserved RFP (Arg-Phe-Pro) motif and they function as anion channels. Bestrophin-4 acts as a calcium-sensitive chloride channel located in the cell membrane. It is believed that Bestrophin-4 also acts as a channel for other physiologically significant anions, such as bicarbonate. Bestrophin-4 is predominantly expressed in the colon, but can be found at low levels in testis, placenta, trachea, spinal chord, lung and retina.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500
Type: Primary
Antigen: BEST4
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge