Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-HAPLN2 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11051R-CY5
BOSSBS-11051R-CY5EA 900 EUR InStock
BOSSBS-11051R-CY5
Anti-HAPLN2 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
HAPLN2 is a 340 amino acid protein encoded by the human gene HAPLN2. HAPLN2 belongs to the HAPLN family and contains one immunoglobulin (Ig)-like, V-type domain and two link domains. HAPLN2 mediates a firm binding of versican V2 to hyaluronic acid. HAPLN2 is believed to play a pivotal role in the formation of the hyaluronan-associated matrix in the central nervous system (CNS), which facilitates neuronal conduction and general structural stabilization. HAPLN2 may also be involved in the formation of extracellular matrices, contributing to perineuronal nets and facilitating the understanding of a functional role of these extracellular matrices. HAPLN2 is found in several nuclei throughout the midbrain and hindbrain in a perineuronal net pattern.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HAPLN2
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge