Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-OTOA/DFNB22 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11060R-A680
BOSSBS-11060R-A680EA 890 EUR InStock
BOSSBS-11060R-A680
Anti-OTOA/DFNB22 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Otoancorin, also known as OTOA, CT108 or DFNB22, is a 1153 amino acid protein belonging to the stereocilin family. Expressed in the inner ear and restricted to the interface between the apical surface of sensory epithelia, otoancorin is suggested to act as an adhesion molecule. Otoancorin ensures the attachment of the inner ear acellular gels to the apical surface of the underlying nonsensory cells. Mutations in the gene encoding otoancorin leads to deafness autosomal recessive type 22 (DFNB22), which is a form of sensorineural hearing loss. Sensorineural deafness results from damage to the neural receptors of the inner ear, the nerve pathways to the brain or the area of the brain that receives sound information. Existing as three alternatively spliced isoforms, otoancorin is encoded by a gene located on human chromosome 16p12.2.

Type: Primary
Antigen: OTOA/DFNB22
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 231-330/1153
Host: Rabbit
Isotype: IgG
Reactivity: Mouse

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge