Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CYTIP Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11062R-HRP
BOSSBS-11062R-HRPEA 900 EUR InStock
BOSSBS-11062R-HRP
Anti-CYTIP Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
CYTIP is a cytoplasmic protein that is involved in lymphocytic cell adhesion. Expressed primarily in hematopoetic cells, CYTIP regulates the activity of cytohesin-1 (an integrin-activating protein involved in cell adhesion) by mediating its recruitment to the leukocyte membrane. Through its ability to bind cytohesin-1, CYTIP is able to sequester it to the cytoplasm, thereby preventing cytohesin-1 translocation to lymphocytes and interrupting the flow of information in the cell adhesion pathway. CYTIP can be recruited from the cytoplasm to the membrane by leukocyte integrins which interact with CYTIP through its PDZ domain. After membrane translocation, CYTIP can be re-located to the cytoplasm via exposure to a phorbol ester. Additionally, CYTIP associates with SNX27 (sorting nexin 27) and helps to coordinate trafficking and signaling complexes. Up-regulation of CYTIP is observed in maturing dendritic cells, suggesting a possible role in developmentally-controlled cell adhesion.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: CYTIP
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge